Learn More
American Research Products Inc Sheep Immunoglobulin A IgA E
Catalog No. 5000328415
Encompass
Sheep Immunoglobulin A (IgA) ELISA Kit from CUSABIO Detection Range 0 234 g/ml-60 g/ml Sensitivity 0 234 g/ml Sample serum plasma Method Competitive
Catalog No. 50-003-28415 Supplier American Research Products Inc Supplier No. CSBE13681SH
Sheep Immunoglobulin A (IgA) ELISA Kit from CUSABIO Detection Range 0 234 g/ml-60 g/ml Sensitivity 0 234 g/ml Sample serum plasma Method Competitive