Learn More
American Research Products Inc Sheep Immunoglobulin M IgM E
Catalog No. 5000328403
Encompass
Sheep Immunoglobulin M (IgM) ELISA Kit from CUSABIO Detection Range 0 156 g/ml-40 g/ml Sensitivity 0 156 g/ml Sample serum plasma Method Competitive
Catalog No. 50-003-28403 Supplier American Research Products Inc Supplier No. CSBE13682SH
Sheep Immunoglobulin M (IgM) ELISA Kit from CUSABIO Detection Range 0 156 g/ml-40 g/ml Sensitivity 0 156 g/ml Sample serum plasma Method Competitive