Learn More
American Research Products Inc Sheep Insulin like growth fact
Catalog No. 5000327184
Encompass
Sheep Insulin-like growth factor II (IGF2) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27184 Supplier American Research Products Inc Supplier No. CSBEL011088SH
Sheep Insulin-like growth factor II (IGF2) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich