Learn More
American Research Products Inc Sheep insulin like growth fact
Catalog No. 5000329290
Encompass
Sheep insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29290 Supplier American Research Products Inc Supplier No. CSBE13753SH
Sheep insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich