Learn More
Biomatik Corporation Sheep Insulin (Ins) Elisa Kit
Supplier:  Biomatik Corporation EKC31515
Encompass_Preferred
Sheep Insulin (Ins) Elisa Kit for detecting Insulin(Ins) in serum, plasma. Species Reactivity: Sheep (Ovis aries). Detection Range: 8-100uIU/ml. Sensitivity: 5uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01318; INS. Target Synonyms: ILPR; IRDN; OTTHUMP00000011162; OTTHUMP00000196038; proinsulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6133