Learn More
American Research Products Inc Sheep Interleukin 1b IL 1b EL
Supplier:  American Research Products Inc CSBE10115SH
Encompass
Sheep Interleukin 1b IL-1b ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29175