Learn More
American Research Products Inc Sheep Interleukin 2 IL 2 ELIS
Catalog No. 5000329660
Encompass
Sheep Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29660 Supplier American Research Products Inc Supplier No. CSBE11217SH
Sheep Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich