Learn More
American Research Products Inc Sheep Interleukin 4 IL 4 ELI
Catalog No. 5000328548
Encompass
Sheep Interleukin 4 (IL-4) ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28548 Supplier American Research Products Inc Supplier No. CSBE15963SH
Sheep Interleukin 4 (IL-4) ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich