Learn More
American Research Products Inc Sheep neutrophil gelatinase as
Catalog No. 5000329469
Encompass
Sheep neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29469 Supplier American Research Products Inc Supplier No. CSBE13392SH
Sheep neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Competitive