Learn More
Biomatik Corporation Sheep Osteocalcin (OC) ELISA Kit
Supplier:  Biomatik Corporation EKU12133
Encompass_Preferred
Sheep Osteocalcin (OC) ELISA Kit for detecting OC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Sheep (Ovis aries). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q1ZZ82. Gene ID: 677670. Target Synonyms: BGLAP; OT; BGP; Bone Gla Protein; Bone Gamma-Carboxyglutamate Protein; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8023