Learn More
Rockland Immunochemicals Sheep Red Blood Cells 100% Washed Pooled Cells 50 mL
SDP
Catalog No. 501943988
Encompass_Preferred
Sheep red blood cells are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents.
Catalog No. 50-194-3988 Supplier Rockland Immunochemicals Supplier No. R4060050
Sheep red blood cells are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents.