Learn More
Carbosynth LLC. Glutaraldehyde-stabilized freshly prepared Sheep Red Blood Cells
SDP
Catalog No. 501253512
Encompass_Preferred
Glutaraldehyde stabilized red blood cells are produced from freshly washed sheep RBCs by brief exposure to a glutaraldehyde-buffer solution and exhaustive washing in saline. This treatment largely preserves the native antigenicity of the erythrocytes while rendering the cells generally resistant to lysis by ostmotic shock, freeze-thawing or immune hemolysis. This reagent can be used directly in hemagglutination procedures, or may be coupled with various proteins for hemagglutination testing.
Catalog No. 50-125-3512 Supplier Carbosynth LLC. Supplier No. 88RS00320ML
Glutaraldehyde stabilized red blood cells are produced from freshly washed sheep RBCs by brief exposure to a glutaraldehyde-buffer solution and exhaustive washing in saline. This treatment largely preserves the native antigenicity of the erythrocytes while rendering the cells generally resistant to lysis by ostmotic shock, freeze-thawing or immune hemolysis. This reagent can be used directly in hemagglutination procedures, or may be coupled with various proteins for hemagglutination testing.