Learn More
Innovative Research Inc Sheep Red Blood Cells Packed 10% 15ml
Catalog No. 502034859
Encompass_Preferred
Sheep Red Blood Cells from Innovative Research are manufactured using whole blood collections from Sheep located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4859 Supplier Innovative Research Inc Supplier No. ISHRBC10P15ML
Sheep Red Blood Cells from Innovative Research are manufactured using whole blood collections from Sheep located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.