Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Sheep Serum, United States Origin
GREENER_CHOICE

Catalog No. MT35042CM Shop All Corning Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT35042CM 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT35042CM Supplier Corning™ Supplier No. 35042CM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Collected at government approved abattoirs in the United States

  • Donor Sheep Serum, United States Origin.
  • Collected at government approved abattoirs in the United States.
  • True pooled and 0.2 micron sterile filtered that passes sterility and mycoplasma.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -40°C to -10°C
Species Ovine
Country of Origin United States
Form Liquid
Quantity 1 L
Serum Treatment None
Sterility Sterile
Source United States Origin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.