Learn More
American Research Products Inc Sheep Transforming growth fact
Supplier:  American Research Products Inc CSBEL023446SH
Encompass
Sheep Transforming growth factor beta-1 (TGFB1) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27789