Learn More
Biomatik Corporation Sheep Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit
Supplier:  Biomatik Corporation EKU11660
Encompass_Preferred
Sheep Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit for detecting TGFb1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Sheep (Ovis aries). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50414. Gene ID: 443417. Target Synonyms: TGF-B1; CED; DPD1; LAP; Camurati-Engelmann Disease; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6290