Learn More
American Research Products Inc Sheep tri iodothyronine T3 E
Catalog No. 5000327407
Encompass
Sheep tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-27407 Supplier American Research Products Inc Supplier No. CSBEQ027510SH
Sheep tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive