Learn More
American Research Products Inc Sheep Tumor Necrosis Factor a
Catalog No. 5000328319
Encompass
Sheep Tumor Necrosis Factor a TNF-a ELISA Kit from CUSABIO Detection Range 3 125 pg/mL-50 pg/mL Sensitivity 3 125 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28319 Supplier American Research Products Inc Supplier No. CSBE13853SH
Sheep Tumor Necrosis Factor a TNF-a ELISA Kit from CUSABIO Detection Range 3 125 pg/mL-50 pg/mL Sensitivity 3 125 pg/mL Sample serum plasma tissue homogenates Method Competitive