Learn More
Research Products International Corp Sheer Lock Ultra-Clear Locking Micro Tubes, 1.5ml Natural, 500 per Package
SDP
Catalog No. 501367508
Encompass
Sheer-Lock micro-tubes offer the security of screw cap tubes at a fraction of the price. Sheer-Locktubes feature a patent pending lock that snaps easily into place with a one hand operation. Lid has a large, rounded design that closes easily and securely without excessive thumb pressure. Additional features include pierceable lid, graduated markings, and frosted writing surfaces. Sheer-Lock micro-tubes are constructed of ultra sheer polypropylene and are autoclavable.
Catalog No. 50-136-7508 Supplier Research Products International Corp Supplier No. 145485
Sheer-Lock micro-tubes offer the security of screw cap tubes at a fraction of the price. Sheer-Locktubes feature a patent pending lock that snaps easily into place with a one hand operation. Lid has a large, rounded design that closes easily and securely without excessive thumb pressure. Additional features include pierceable lid, graduated markings, and frosted writing surfaces. Sheer-Lock micro-tubes are constructed of ultra sheer polypropylene and are autoclavable.