Learn More
Med Vet International Equispon Absorbable Get Sheets, 125mm x 80mm x 10mm, J1275, Each
Catalog No. 5000495646
Encompass_Preferred
A local hemostatic sponge for use in surgical procedures that involve venous bleeding and oozing where standard hemostats are difficult or impractical• Achieves hemostatis rapidly in a few minutes. • Can be trimmed to size or sutured in place• Absorbs 50 times its own weight• Uniform porosity• Can use with medications and not affect its hemostatic properties• Sterile double packaged120mm x 80mm x 10mm
Catalog No. 50-004-95646 Supplier Med Vet International Supplier No. JOR1275
A local hemostatic sponge for use in surgical procedures that involve venous bleeding and oozing where standard hemostats are difficult or impractical• Achieves hemostatis rapidly in a few minutes. • Can be trimmed to size or sutured in place• Absorbs 50 times its own weight• Uniform porosity• Can use with medications and not affect its hemostatic properties• Sterile double packaged120mm x 80mm x 10mm