Learn More
Lakeside Manufacturing Inc Mobile Sheet Pan Drying Rack, Channel Slides, Fits 14 x 18-in. or 18 x 26-in. Pans, 120 Pan Capacity
Catalog No. 502884441
Encompass_Preferred
Conveniently store domes and trays in these open racks for efficient drying. The Lakeside 848 Mobile Sheet Pan Drying Rack, with channel slides, fits 14 x 18-in. or 18 x 26-in. pans, and has a 120 pan capacity. 11- gauge all welded stainless steel base ensures superior strength and balance. This unit has a 14-gauge stainless steel frame and “L” supports increase stability to prevent accidents and spills. Rugged all-welded frame of stainless steel square tubing. Accommodates 18" x 26" and 14" x 18" pans or trays. 5" (127) corner stem no-mark polyurethane casters offer flexibility to move rack from sink to storage area. Stainless steel channel slides with 1½" spacing. Solid stainless steel tray shelves. The pan-stop ledge prevents pans from sliding off of back side. Nobody beats Lakeside's lead times. Made in the USA.
Catalog No. 50-288-4441 Supplier Lakeside Manufacturing Inc Supplier No. 848
Add to Cart
Add to Cart
Request a Quote
Conveniently store domes and trays in these open racks for efficient drying. The Lakeside 848 Mobile Sheet Pan Drying Rack, with channel slides, fits 14 x 18-in. or 18 x 26-in. pans, and has a 120 pan capacity. 11- gauge all welded stainless steel base ensures superior strength and balance. This unit has a 14-gauge stainless steel frame and “L” supports increase stability to prevent accidents and spills. Rugged all-welded frame of stainless steel square tubing. Accommodates 18" x 26" and 14" x 18" pans or trays. 5" (127) corner stem no-mark polyurethane casters offer flexibility to move rack from sink to storage area. Stainless steel channel slides with 1½" spacing. Solid stainless steel tray shelves. The pan-stop ledge prevents pans from sliding off of back side. Nobody beats Lakeside's lead times. Made in the USA.