Learn More
MSC Mag-Mate Magnetic Sheet Separator, Heavy-Duty, 7 to 12 Gauge, 1 ea
Catalog No. 5000511134
Encompass
This heavy-duty magnetic sheet separator is designed to efficiently separate oily sheets for safer, faster handling in metalworking operations. The fanner uses a powerful ceramic magnet to create a gap between sheets, enabling easier manual or automated feeding. It is built for durability and flexibility in industrial environments.
  • Separates metal sheets ranging from 7 to 12 gauge
  • Equipped with a powerful ceramic magnet
  • Features durable, welded steel construction
  • Includes wear strips on the magnetic face for extended service life
  • Supplied with a bolt-on angle base plate and handle with mounting hardware
  • Supports operating temperatures from -148°F to 480°F
  • Pre-tapped holes provided for secure mounting to equipment
Catalog No. 50-005-11134 Supplier MSC Supplier No. 06586085
This heavy-duty magnetic sheet separator is designed to efficiently separate oily sheets for safer, faster handling in metalworking operations. The fanner uses a powerful ceramic magnet to create a gap between sheets, enabling easier manual or automated feeding. It is built for durability and flexibility in industrial environments.
  • Separates metal sheets ranging from 7 to 12 gauge
  • Equipped with a powerful ceramic magnet
  • Features durable, welded steel construction
  • Includes wear strips on the magnetic face for extended service life
  • Supplied with a bolt-on angle base plate and handle with mounting hardware
  • Supports operating temperatures from -148°F to 480°F
  • Pre-tapped holes provided for secure mounting to equipment