Learn More
Grainger Sheet SS 4 in Overall W PK6
Catalog No. 502764478
Encompass_Preferred
The 16NH24 is a 16 ton hydraulic bottle jack manufactured by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for strength and stability. It is designed for use with hydraulic fluid and has a built-in bypass device. The jack has a two piece handle for ease of pumping and can lift up to 32,000 pounds. Its compact design makes it portable and suitable for use in auto shops and construction sites where heavy loads need to be lifted.
Catalog No. 50-276-4478 Supplier Grainger Supplier No. 16NH24
The 16NH24 is a 16 ton hydraulic bottle jack manufactured by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for strength and stability. It is designed for use with hydraulic fluid and has a built-in bypass device. The jack has a two piece handle for ease of pumping and can lift up to 32,000 pounds. Its compact design makes it portable and suitable for use in auto shops and construction sites where heavy loads need to be lifted.