Learn More
Grainger Sheet Tin 10 in Steel Plated PK6
Catalog No. 502764587
Encompass_Preferred
The Grainger 16NH23 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and protected from dirt and moisture. This high quality bottle jack is designed for shop use and farm equipment lifting. With a 16 ton capacity, it can safely lift up to 32,000 pounds. The jack weighs 43 pounds and measures 11 x 7 x 8 inches.
Catalog No. 50-276-4587 Supplier Grainger Supplier No. 16NH23
The Grainger 16NH23 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and protected from dirt and moisture. This high quality bottle jack is designed for shop use and farm equipment lifting. With a 16 ton capacity, it can safely lift up to 32,000 pounds. The jack weighs 43 pounds and measures 11 x 7 x 8 inches.