Learn More
KERRY INGREDIENTS AND FLAVOURS SHEFF-CHO PLUS PF ACF,5KG PAIL
Catalog No. NC2282842
Encompass
Sheff-CHO Plus PF ACF is a protein –free version of Sheff-CHO ACF which contains no intact proteins for those with concerns regarding downstream purification.
This item is not returnable. View return policy
Catalog No. NC2282842 Supplier KERRY INGREDIENTS AND FLAVOURS Supplier No. 20600740
Sheff-CHO Plus PF ACF is a protein –free version of Sheff-CHO ACF which contains no intact proteins for those with concerns regarding downstream purification.