Learn More
Enterprise Technology Solutions Sheila Shine Inc. Cleaner/Polish, 1 Gallon, 4/CT, Blue/White
SDP
Catalog No. 5000790043
Encompass_Preferred
Stainless steel polish/surface preservative features an easy-to-use design that requires minimal rubbing and polishing to reduce maintenance times. Versatile formula removes greasy film and water marks, resists fingerprints and streaking, repels water and preserves surfaces against deterioration. It even prevents liquid penetration, eliminates streaking and resists finger marks. Polish and surface preservative is perfect for stainless steel, chrome, aluminum, brass, formica, porcelain, fiberglass, enamel, leather and more.
Catalog No. 50-007-90043 Supplier Enterprise Technology Solutions Supplier No. SSISSCA128CT
Stainless steel polish/surface preservative features an easy-to-use design that requires minimal rubbing and polishing to reduce maintenance times. Versatile formula removes greasy film and water marks, resists fingerprints and streaking, repels water and preserves surfaces against deterioration. It even prevents liquid penetration, eliminates streaking and resists finger marks. Polish and surface preservative is perfect for stainless steel, chrome, aluminum, brass, formica, porcelain, fiberglass, enamel, leather and more.