Learn More
Sheldon Manufacturing Inc SHEL LAB CO2 WJ 5CUFT INCUB
SDP
Catalog No. NC1263810
Encompass_Preferred
CO2 INCUBATOR, WATER JACKET, 5 CU FT, IR, HEPA, CO2 TANK SWITCH, 115V. Model SCO5W water jacketed CO2 incubator is an ideal unit for cell culture applications. It provides a precisely controlled, accurate, pure environment throughout the culturing process. Shel Lab CO2 incubators are designed to provide the uniform temperatures and highly stable CO2 environments that are necessary to prevent shocks to cells. Also available in a benchtop 1.5 cu.ft./42 L size (Model SCO2W), and SCO5W units can be stacked with the use of a stacking rack (PN 99751223). Includes Dual beam IR sensor and RH sensor as standard. More information available at www.shellab.com
Catalog No. NC1263810 Supplier Sheldon Manufacturing Inc Supplier No. SCO5W
Add to Cart
Add to Cart
Request a Quote
CO2 INCUBATOR, WATER JACKET, 5 CU FT, IR, HEPA, CO2 TANK SWITCH, 115V. Model SCO5W water jacketed CO2 incubator is an ideal unit for cell culture applications. It provides a precisely controlled, accurate, pure environment throughout the culturing process. Shel Lab CO2 incubators are designed to provide the uniform temperatures and highly stable CO2 environments that are necessary to prevent shocks to cells. Also available in a benchtop 1.5 cu.ft./42 L size (Model SCO2W), and SCO5W units can be stacked with the use of a stacking rack (PN 99751223). Includes Dual beam IR sensor and RH sensor as standard. More information available at www.shellab.com