Learn More
Shelby™ Specialty 5280 Gloves
Catalog No. 19167783 Shop All Shelby Specialty Gloves Products
Click to view available options
Size:
Medium
Catalog No. Size
19-167-783 Medium
1 options
This item is not returnable. View return policy
Catalog No. 19-167-783 Supplier Shelby™ Supplier No. 5280GM
This item is not returnable. View return policy
Specialty thermal gloves are resistant to flames, heat, and punctures for fire management or other industrial applications.
  • Heavyweight, gauntlet-style gloves
  • Grain elkhide leather along with Shelby abrasion-resistant brushed pigskin leather
  • Shelby Black Line Nomex™ 101.6 mm (4 in.) wristlet
  • GORE RT7100 glove barrier fabric
  • Gunn-cut pattern with wing thumb and wrap-around index finger
Certifications
SEI Certified
Specifications
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Color Tan
Material Pigskin Leather, Elkskin Leather
Product Type Structural Firefighting Gloves
Size Medium
Glove Cuff Style Gauntlet
Liner Material Thermal