Learn More
Shelby™ Structural Direct Grip Gloves
Catalog No. 19147320 Shop All Shelby Specialty Gloves Products
Click to view available options
Material:
Aramid Fiber, Leather
Catalog No. Material
19-147-320 Aramid Fiber, Leather
1 options
Catalog No. 19-147-320 Supplier Shelby™ Supplier No. 5285J
Innovative wash-and-wear palm stays soft and supple even after repeated washing
  • NFPA-compliant structural gloves provide superior dexterity, grip, and thermal protection
  • Highly engineered insular construction acts as both a thermal and flame barrier
  • Breathable, CrossTech™ Direct Gripilm technology insert permanently attached to Aramid Fiber tech fleece
  • Fingertips are covered with reflective 3M™ Scotchlite™
  • 3.5 oz. Koala-tanned leather palm
Specifications
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Material Aramid Fiber, Leather
Product Type Structural Firefighting Gloves
Size 2X-Large
Glove Cuff Style Gauntlet
Liner Material Thermal