Learn More
Sheldon Manufacturing Inc SHELDON 3.1 HRZ AIR FLOW OVEN
SDP
Catalog No. NC0683287
Encompass_Preferred
OVEN, FORCED AIR, DIGITAL WITH TIMER, Horizontal Air Flow, 3 CUFT, 115V. The SMO3 has a quick ramp-up time to 306°C for faster cycle times. This unit includes an advanced PID temperature control system for sensitive response. These ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing. This unit is equipped with a stainless steel interior for long life operation and easy cleaning. Its triple wall construction minimizes external wall temperature which meets CE requirements to keep workspaces cool. The SMO3’s forced air ovens deliver precise uniformity, air distribution and the peace of mind expected of the SHEL LAB brand. More information available at www.shellab.com
Catalog No. NC0683287 Supplier Sheldon Manufacturing Inc Supplier No. SMO3
OVEN, FORCED AIR, DIGITAL WITH TIMER, Horizontal Air Flow, 3 CUFT, 115V. The SMO3 has a quick ramp-up time to 306°C for faster cycle times. This unit includes an advanced PID temperature control system for sensitive response. These ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing. This unit is equipped with a stainless steel interior for long life operation and easy cleaning. Its triple wall construction minimizes external wall temperature which meets CE requirements to keep workspaces cool. The SMO3’s forced air ovens deliver precise uniformity, air distribution and the peace of mind expected of the SHEL LAB brand. More information available at www.shellab.com