Learn More
Grainger New Age Industrial Solid Shelf, Aluminum, 36 in. x 18 in., 1 ea.
Catalog No. 502785445
Encompass_Preferred
This 36 in. x 18 in. solid adjustable shelf is designed for versatile and hygienic storage. Constructed from 18 gauge aluminum, the shelf features a durable, smooth surface that is ideal for organizing smaller items and preventing spills. It is equipped with reinforced support tubing and welded corner collars for stability, making it a reliable solution for various storage environments.
  • NSF certified for hygiene and safety
  • Weight capacity of 400 lb. per shelf
  • Raised 1/8 in. marine edge to contain spills and simplify cleaning
  • Adjustable design compatible with standard shelving poles
  • Construction includes two 1.5 in. x 1.5 in. x 0.070 in. wall support tubes
  • Backed by a one-year guarantee against workmanship and material defects, plus a lifetime guarantee against rust and corrosion
Catalog No. 50-278-5445 Supplier Grainger Supplier No. 18K918
This 36 in. x 18 in. solid adjustable shelf is designed for versatile and hygienic storage. Constructed from 18 gauge aluminum, the shelf features a durable, smooth surface that is ideal for organizing smaller items and preventing spills. It is equipped with reinforced support tubing and welded corner collars for stability, making it a reliable solution for various storage environments.
  • NSF certified for hygiene and safety
  • Weight capacity of 400 lb. per shelf
  • Raised 1/8 in. marine edge to contain spills and simplify cleaning
  • Adjustable design compatible with standard shelving poles
  • Construction includes two 1.5 in. x 1.5 in. x 0.070 in. wall support tubes
  • Backed by a one-year guarantee against workmanship and material defects, plus a lifetime guarantee against rust and corrosion