Learn More
Grainger Shelf Bin, Blue, 7 in H x 20 1/2 in L x 11 in W, each
Catalog No. NC1981735
Encompass_Preferred
Designed with strong, durable polypropylene construction, this stackable, waterproof shelf bin is rust and corrosion resistant. Autoclavable up to 250°F, it's also resistant to extreme cold. The raised front opening maximizes storage capacity. The large, molded-in front slot holds ID labels.
Catalog No. NC1981735 Supplier Grainger Supplier No. 24K184
Designed with strong, durable polypropylene construction, this stackable, waterproof shelf bin is rust and corrosion resistant. Autoclavable up to 250°F, it's also resistant to extreme cold. The raised front opening maximizes storage capacity. The large, molded-in front slot holds ID labels.