Learn More
Uline Plastic Shelf Bins - 7 x 12 x 4, Black, 20
Catalog No. NC2049746
Encompass
Long-lasting storage for small parts and components, Nest to save storage space, Molded label holder to identify contents, Rear notch allows bins to slide and hang open on Shelf Bin Organizers, FDA compliant resins
Catalog No. NC2049746 Supplier Uline Supplier No. S13397BLCS
Long-lasting storage for small parts and components, Nest to save storage space, Molded label holder to identify contents, Rear notch allows bins to slide and hang open on Shelf Bin Organizers, FDA compliant resins