Learn More
Grainger Shelf Cap 1 000 lb 60 x 30 in
Catalog No. 502752970
Encompass_Preferred
The Grainger 14C738 is a Westinghouse 15 Amp, 125 Volt, 3-Wire Grounding Plug with Straight Blades and Side Wire Holes. It has side wire holes that allow for easy installation and removal. This plug features heavy duty construction with high impact resistant nylon housing. It can be used on extension cords and power tools. The plug has a NEMA configuration of 5-15P. It is UL listed and CSA certified. The plug is 15 amps, 125 volts with a straight blade design.
Catalog No. 50-275-2970 Supplier Grainger Supplier No. 14C738
The Grainger 14C738 is a Westinghouse 15 Amp, 125 Volt, 3-Wire Grounding Plug with Straight Blades and Side Wire Holes. It has side wire holes that allow for easy installation and removal. This plug features heavy duty construction with high impact resistant nylon housing. It can be used on extension cords and power tools. The plug has a NEMA configuration of 5-15P. It is UL listed and CSA certified. The plug is 15 amps, 125 volts with a straight blade design.