Learn More
Grainger Shelf Cap 1 200 lb 60 x 48 in
Catalog No. 502753374
Encompass_Preferred
The Grainger 14L133 is a Westward black nitrile coated work glove with a textured grip. It has a 13 gauge seamless knit liner for comfort and durability. The nitrile coating provides oil and water resistance. The gloves are sold in pairs, ambidextrous with no specific left or right hand. They come in sizes S-2XL.
Catalog No. 50-275-3374 Supplier Grainger Supplier No. 14L133
The Grainger 14L133 is a Westward black nitrile coated work glove with a textured grip. It has a 13 gauge seamless knit liner for comfort and durability. The nitrile coating provides oil and water resistance. The gloves are sold in pairs, ambidextrous with no specific left or right hand. They come in sizes S-2XL.