Learn More
Grainger Shelf Cap 500 lb 48 x 36 in
Catalog No. 502751740
Encompass_Preferred
The Grainger Freestanding Wire Shelving is made of steel with a chrome finish. It has 4 adjustable shelves that can hold up to 800 lbs per shelf. The shelves are 30" deep x 48" wide. The unit is available in 36", 48", 60", and 72" heights. Assembly required. Weight capacity 3,200 lbs total. NSF certified. Ideal for commercial and industrial use. Provides open storage for easy access.
Catalog No. 50-275-1740 Supplier Grainger Supplier No. 14C734
The Grainger Freestanding Wire Shelving is made of steel with a chrome finish. It has 4 adjustable shelves that can hold up to 800 lbs per shelf. The shelves are 30" deep x 48" wide. The unit is available in 36", 48", 60", and 72" heights. Assembly required. Weight capacity 3,200 lbs total. NSF certified. Ideal for commercial and industrial use. Provides open storage for easy access.