Learn More
Grainger Shelf Cap 700 lb 48 x 24 in
Catalog No. 502749933
Encompass_Preferred
The Grainger Approved Shelf Cap is a plastic shelf cap designed to protect the edges of shelving. It measures 4" long x 1" wide x 1/4" thick and has a black finish. The cap slips over the front edge of metal shelving up to 13 gauge to provide a finished look and help prevent injury from sharp edges. It can be used on wall standards, wall brackets, slotted angle and other metal shelving. The shelf cap is made of durable ABS plastic for long life.
Catalog No. 50-274-9933 Supplier Grainger Supplier No. 14L185
The Grainger Approved Shelf Cap is a plastic shelf cap designed to protect the edges of shelving. It measures 4" long x 1" wide x 1/4" thick and has a black finish. The cap slips over the front edge of metal shelving up to 13 gauge to provide a finished look and help prevent injury from sharp edges. It can be used on wall standards, wall brackets, slotted angle and other metal shelving. The shelf cap is made of durable ABS plastic for long life.