Learn More
Poltex Shelf System Drawer - 14 x 17 x 7 (in.)
SDP
Catalog No. 22022536
Change view
Click to view available options
Clinical Lab Organizer
Catalog No.
22-022-536 Clinical Lab Organizer
1 options
Catalog No. 22-022-536 Supplier Poltex Supplier No. SHLFSYSDWR7
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This drawer unit is part of the Poltex shelf organization system. All products in the Poltex shelf organization system line can stack on one another using the foot and pocket design to create the perfect shelf storage set up.
  • Drawer unit
  • Super durable and cleanable PETG construction
  • Rubber bumpons on base for non-slip placement
  • All products in the Poltex Shelf Organization System line can stack on one another using our foot and pocket design to create the perfect shelf storage set up
Specifications
Product Type Clinical Lab Organizer
No. of Doors None
Color Clear
No. of Shelves None
Autoclavable Not Autoclavable
Material PETG