Learn More
Andwin Scientific SHELL VIAL 1 DRAM 144/PK
SDP
Catalog No. 50949345
Encompass_Preferred
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage. The vials come with polyethylene open bottom snap-caps, Titeseal, which are packed separately. Specimen Preparation and Embedding Supplies: Vials, Racks, Holders, and Lyophilization Containers: Vials; Shell: Clear Glass, Plain Top: 1 dr. (3.7ml) Capacity, 15 OD mm, 45 Hgt mm, 144/pk
Catalog No. 50-949-345 Supplier Andwin Scientific Supplier No. 7263110
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage. The vials come with polyethylene open bottom snap-caps, Titeseal, which are packed separately. Specimen Preparation and Embedding Supplies: Vials, Racks, Holders, and Lyophilization Containers: Vials; Shell: Clear Glass, Plain Top: 1 dr. (3.7ml) Capacity, 15 OD mm, 45 Hgt mm, 144/pk