Learn More
Electron Microscopy Sciences Shell Vial Holder
SDP
Catalog No. 50949304
Encompass_Preferred
Machined from solid aluminum, holds 5 (#72631-10) shell vials. It is ideal for handling cold or hot samples for prolonged periods of time out of the refrigerator or oven. Comes complete with a handle. Measurements: 4 3/4" x 1 1/4" x 3/4".
Catalog No. 50-949-304 Supplier Electron Microscopy Sciences Supplier No. 72640
Machined from solid aluminum, holds 5 (#72631-10) shell vials. It is ideal for handling cold or hot samples for prolonged periods of time out of the refrigerator or oven. Comes complete with a handle. Measurements: 4 3/4" x 1 1/4" x 3/4".