Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Shell Vial Kit

Catalog No. 01672358 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-672-358 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-358
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vial constructed of clear glass and equipped with a polyethylene plug.

  • 2 mL shell vial
  • Clear class with polyethylene plug
  • Pack of 100
  • 31.5 mm x 11.6 mm dimensions.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Diameter (Metric) Outer 31.5 mm
Material Polyethylene
Sterility Non-sterile
Disposable Single-use
Volume (Metric) 2 mL
Closure Type Plug
Quantity 100/Pk.
Fit for Purpose Certified Quality
Vial Type Glass
Product Type Plug
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.