Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America™ Shelter in Place Essentials Emergency Kit, 10 Pack
SDP

Catalog No. 19777022
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-777-022 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-777-022 Supplier Ready America™ Supplier No. 70506
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready America™ Shelter in Place Essentials Emergency Kit is convenient and designed to grab for a quick escape or to shelter in place.

  • Includes the following:
    • 1 Food Bar (2400 Calorie) (5-year shelf life)
    • 6 Water Pouches 4.225 oz. (5-year shelf life)
    • 1 Emergency Lightstick (12-hr)
    • 1 Emergency Whistle
    • 1 Survival Blanket
    • 1 Emergency Poncho
    • 1 Pair of Hand Warmers
    • 1 Pocket Tissue Pack
    • 1 Toothbrush
    • 1 Toothpaste
    • 3 Pre-Moistened Towelettes
    • 1 Bio-Hazard Bag
    • 3 Antiseptic Cleansing Wipes
    • 1 Triple Antibiotic Ointment Pack
    • 1 Ibuprofen (200 mg)
    • 1 Butterfly Wound Closure
    • 5 Medium Bandages
    • 5 Junior Bandage (0.2 x 0.5 in.)
    • 1 First-Aid Guide

Specifications

Product Type Emergency Supplies
Color Clear
Material Plastic
Height (English) 12 in.
Height (Metric) 30.48 cm
Length (English) 12 in.
Length (Metric) 30.48 cm
Weight (English) 13.2 lb.
Weight (Metric) 6 kg
Width (English) 20 in.
Width (Metric) 50.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.