Learn More
Fisherbrand™ Isotemp™ Shelf
GSA_VA
Catalog No. 150145851 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
16 x 22 in.
Catalog No. Dimensions (L x W)
15-014-5851 16 x 22 in.
1 options
Catalog No. 15-014-5851
Add to Cart
Add to Cart
Material: Wire; For Use With: Isotemp gravity convection oven/Incubator, 6.2/6.8 cu. ft.
  • Material: Stainless steel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Isotemp gravity convection oven/incubator, 6.2/6.8 cu. ft.
Material Wire
Type Shelf
Dimensions (L x W) 16 x 22 in.