Learn More
Grainger JAMCO Storage Cabinet, Steel, 48 in x 24 in x 54 in, 2 Shelves, 1 ea
Catalog No. 502787113
Encompass_Preferred
This industrial stationary storage cabinet is designed for use in demanding environments such as machine shops, plants, and warehouses. Constructed from 12 ga steel with a durable powder-coated finish, this fully welded unit provides high-impact resistance and security. The cabinet features a ventilated body and door design, offering improved airflow and visibility.
  • Features a super duty construction with 12 ga steel panels
  • Includes two fixed shelves with a load capacity of 1,500 lbs. per shelf
  • Equipped with two doors and a swing handle with a padlock hasp for security
  • Designed as a fully welded and assembled stationary unit with legs
  • Overall dimensions of 48 in. W x 24 in. D x 54 in. H
Catalog No. 50-278-7113 Supplier Grainger Supplier No. 18G988
This industrial stationary storage cabinet is designed for use in demanding environments such as machine shops, plants, and warehouses. Constructed from 12 ga steel with a durable powder-coated finish, this fully welded unit provides high-impact resistance and security. The cabinet features a ventilated body and door design, offering improved airflow and visibility.
  • Features a super duty construction with 12 ga steel panels
  • Includes two fixed shelves with a load capacity of 1,500 lbs. per shelf
  • Equipped with two doors and a swing handle with a padlock hasp for security
  • Designed as a fully welded and assembled stationary unit with legs
  • Overall dimensions of 48 in. W x 24 in. D x 54 in. H