Learn More
Grainger Jamco Storage Cabinet, Steel, 54 in. x 60 in., 4 Shelves, 1 ea.
Catalog No. 502787108
Encompass_Preferred
This super-duty storage cabinet is designed for secure and organized storage in industrial environments. Featuring a robust steel construction with a durable powder-coated finish, it provides a stable and reliable solution for heavy-duty applications.
  • Constructed from 12 ga steel for maximum durability
  • Equipped with 4 shelves, each supporting up to 1,500 lb
  • Includes 2 ventilated doors with a padlock hasp for security
  • Stationary base with legs for stable placement
  • Fully assembled for immediate installation
  • Overall dimensions are 54 in. H x 60 in. W x 36 in. D
Catalog No. 50-278-7108 Supplier Grainger Supplier No. 18H008
This super-duty storage cabinet is designed for secure and organized storage in industrial environments. Featuring a robust steel construction with a durable powder-coated finish, it provides a stable and reliable solution for heavy-duty applications.
  • Constructed from 12 ga steel for maximum durability
  • Equipped with 4 shelves, each supporting up to 1,500 lb
  • Includes 2 ventilated doors with a padlock hasp for security
  • Stationary base with legs for stable placement
  • Fully assembled for immediate installation
  • Overall dimensions are 54 in. H x 60 in. W x 36 in. D