Learn More
Grainger Jamco Ventilated Storage Cabinet, Steel, 60 in. x 54 in. x 36 in., 1 ea.
Catalog No. 502787738
Encompass_Preferred
This super-duty industrial storage cabinet is constructed from 12 ga. steel with a durable black powder coated finish, providing a robust solution for demanding environments. Designed for stationary use, this cabinet features two shelves and two ventilated swing doors for secure, organized storage.
  • Features 12 ga. steel construction with powder coating for enhanced durability
  • Equipped with two sturdy shelves each supporting up to 1,500 lbs.
  • Includes two ventilated swing doors for increased airflow
  • Secured with a padlock hasp for reliable locking
  • Measures 152.4 cm (60 in.) wide by 137.16 cm (54 in.) high by 91.44 cm (36 in.) deep
Catalog No. 50-278-7738 Supplier Grainger Supplier No. 18G993
This super-duty industrial storage cabinet is constructed from 12 ga. steel with a durable black powder coated finish, providing a robust solution for demanding environments. Designed for stationary use, this cabinet features two shelves and two ventilated swing doors for secure, organized storage.
  • Features 12 ga. steel construction with powder coating for enhanced durability
  • Equipped with two sturdy shelves each supporting up to 1,500 lbs.
  • Includes two ventilated swing doors for increased airflow
  • Secured with a padlock hasp for reliable locking
  • Measures 152.4 cm (60 in.) wide by 137.16 cm (54 in.) high by 91.44 cm (36 in.) deep