Learn More
Grainger JAMCO Storage Cabinet, Steel, Ventilated, 3 Fixed Shelves, 1 ea
Catalog No. 502787112
Encompass_Preferred
This industrial stationary storage cabinet is designed for secure, ventilated storage of tools, parts, and equipment in demanding environments such as warehouses, workshops, and manufacturing facilities. The cabinet features a fully welded, pre-assembled steel construction with a durable powder-coated finish to withstand heavy-duty use. Its ventilated body panels promote airflow, while the freestanding design includes stable legs and a secure locking mechanism for organized storage.
  • Features 3 fixed shelves with a load capacity of 1,500 lb per shelf
  • Constructed from 12 ga steel shelves and 14 ga steel panels
  • Includes a swing handle with a padlock hasp for secure closure
  • Fully welded and assembled frame for immediate use
  • Overall load capacity of 3,000 lb
  • Dimensions: 72 in. W x 54 in. H x 36 in. D
Catalog No. 50-278-7112 Supplier Grainger Supplier No. 18H002
This industrial stationary storage cabinet is designed for secure, ventilated storage of tools, parts, and equipment in demanding environments such as warehouses, workshops, and manufacturing facilities. The cabinet features a fully welded, pre-assembled steel construction with a durable powder-coated finish to withstand heavy-duty use. Its ventilated body panels promote airflow, while the freestanding design includes stable legs and a secure locking mechanism for organized storage.
  • Features 3 fixed shelves with a load capacity of 1,500 lb per shelf
  • Constructed from 12 ga steel shelves and 14 ga steel panels
  • Includes a swing handle with a padlock hasp for secure closure
  • Fully welded and assembled frame for immediate use
  • Overall load capacity of 3,000 lb
  • Dimensions: 72 in. W x 54 in. H x 36 in. D