Learn More
MSC Shepherd Stem Mount Swivel Caster, 3 in Diameter, 15/16 in Wide, 3-7/8 in OAH
Catalog No. 5000534486
Encompass
This stem mount swivel caster is designed for various applications, particularly in electronic and business equipment. It features a durable construction with a chrome finish and offers smooth mobility with its precision ball bearing.
  • Made from durable neoprene and rubber materials.
  • Features a capacity of 110 lb.
  • Equipped with a precision ball bearing for smooth operation.
  • Designed with a friction stem for secure mounting.
  • Has a 3 in wheel diameter.
  • Constructed from stamp formed components.
  • Finished with a chrome coating.
  • Suitable for electronic and business equipment.
  • Offers swivel functionality.
  • Country of origin: China.
Catalog No. 50-005-34486 Supplier MSC Supplier No. 74582800
This stem mount swivel caster is designed for various applications, particularly in electronic and business equipment. It features a durable construction with a chrome finish and offers smooth mobility with its precision ball bearing.
  • Made from durable neoprene and rubber materials.
  • Features a capacity of 110 lb.
  • Equipped with a precision ball bearing for smooth operation.
  • Designed with a friction stem for secure mounting.
  • Has a 3 in wheel diameter.
  • Constructed from stamp formed components.
  • Finished with a chrome coating.
  • Suitable for electronic and business equipment.
  • Offers swivel functionality.
  • Country of origin: China.