Learn More
MSC Shepherd Swivel Caster, Neoprene, 3 in Diameter, 110 lb Capacity
Catalog No. 5000513088
Encompass
This swivel caster is designed for electronic and business equipment applications. It features a 3-inch diameter neoprene wheel with a 110 lb load capacity, a precision ball bearing, and a 7/16 x 7/8 inch friction stem.
  • Neoprene wheel material
  • 110 lb load capacity
  • Precision ball bearing
  • Friction stem type
  • 7/16 x 7/8 in stem dimensions
  • Swivel functionality
  • 3 in wheel diameter
  • Stamp formed construction
  • Chrome finish
  • Suitable for electronic and business equipment
Catalog No. 50-005-13088 Supplier MSC Supplier No. 74582875
This swivel caster is designed for electronic and business equipment applications. It features a 3-inch diameter neoprene wheel with a 110 lb load capacity, a precision ball bearing, and a 7/16 x 7/8 inch friction stem.
  • Neoprene wheel material
  • 110 lb load capacity
  • Precision ball bearing
  • Friction stem type
  • 7/16 x 7/8 in stem dimensions
  • Swivel functionality
  • 3 in wheel diameter
  • Stamp formed construction
  • Chrome finish
  • Suitable for electronic and business equipment