Learn More
MSC Shepherd Swivel Top Plate Caster, Polyurethane, 5 in Wheel Dia, 225 lb Capacity
Catalog No. 5000533190
Encompass
This Shepherd Swivel Top Plate Caster features a durable polyurethane wheel with a 5-inch diameter and 1-1/4 inch width, designed to support a load capacity of up to 225 pounds. Its hardened-heat treated construction and zinc finish ensure reliability for institutional applications.
  • Polyurethane wheel
  • Ball bearing type
  • Hardened-heat treated construction
  • Zinc finish/coating
  • Designed for institutional applications
  • Swivel style
Catalog No. 50-005-33190 Supplier MSC Supplier No. 06465645
This Shepherd Swivel Top Plate Caster features a durable polyurethane wheel with a 5-inch diameter and 1-1/4 inch width, designed to support a load capacity of up to 225 pounds. Its hardened-heat treated construction and zinc finish ensure reliability for institutional applications.
  • Polyurethane wheel
  • Ball bearing type
  • Hardened-heat treated construction
  • Zinc finish/coating
  • Designed for institutional applications
  • Swivel style